Learn More
Med Vet International Super Absorbent Dressing HydraLock™ SA 6 X 10 Inch Rectangle, 60610, 10/Box
Catalog No. 503043077
Encompass_Preferred
Super absorbent core for moderate to highly exudating wounds Nonadherent contact layer will not stick to wounds Waterproof backing prevents strikethrough Assists with moist wound healing and autolysis Minimizes the risk of maceration Supports formation of granulation tissue and epithelialization
Catalog No. 50-304-3077 Supplier Med Vet International Supplier No. 949203BX
Super absorbent core for moderate to highly exudating wounds Nonadherent contact layer will not stick to wounds Waterproof backing prevents strikethrough Assists with moist wound healing and autolysis Minimizes the risk of maceration Supports formation of granulation tissue and epithelialization