Learn More
Med Vet International McKesson Adhesive Foam Dressing with Border, 3 x 3 Inch, 16-4670, Each
Catalog No. 503044493
Encompass_Preferred
Foam Dressing McKesson 3 X 3 Inch Square Acrylic Adhesive with Border Sterile Highly conformable and remains intact on skin Skin-friendly acrylic adhesive ensures that the dressing remains secure May be used as dressing under compression wraps or Unna boots Affixes securely to dry, intact skin; seals the wound margins to help prevent exudate from leaking on surrounding tissue, minimizing risk of maceration Breathable, yet moisture and bacteria resistant outer film layer Absorbent inner hydrocellular foam pad Dressing may be lifted and repositioned without losing its adherent properties Maintains a moist wound environment to help promote better healing
Catalog No. 50-304-4493 Supplier Med Vet International Supplier No. 1138279EA
Foam Dressing McKesson 3 X 3 Inch Square Acrylic Adhesive with Border Sterile Highly conformable and remains intact on skin Skin-friendly acrylic adhesive ensures that the dressing remains secure May be used as dressing under compression wraps or Unna boots Affixes securely to dry, intact skin; seals the wound margins to help prevent exudate from leaking on surrounding tissue, minimizing risk of maceration Breathable, yet moisture and bacteria resistant outer film layer Absorbent inner hydrocellular foam pad Dressing may be lifted and repositioned without losing its adherent properties Maintains a moist wound environment to help promote better healing