Learn More
Med Vet International McKesson Adhesive Foam Dressing with Border, 3 x 3 Inch, 16-4670, 10/Box
Catalog No. 503043269
Encompass_Preferred
Foam Dressing McKesson 3 X 3 Inch Square Acrylic Adhesive with Border Sterile Highly conformable and remains intact on skin Skin-friendly acrylic adhesive ensures that the dressing remains secure May be used as dressing under compression wraps or Unna boots Affixes securely to dry, intact skin; seals the wound margins to help prevent exudate from leaking on surrounding tissue, minimizing risk of maceration Breathable, yet moisture and bacteria resistant outer film layer Absorbent inner hydrocellular foam pad Dressing may be lifted and repositioned without losing its adherent properties Maintains a moist wound environment to help promote better healing
Catalog No. 50-304-3269 Supplier Med Vet International Supplier No. 1138279BX
Foam Dressing McKesson 3 X 3 Inch Square Acrylic Adhesive with Border Sterile Highly conformable and remains intact on skin Skin-friendly acrylic adhesive ensures that the dressing remains secure May be used as dressing under compression wraps or Unna boots Affixes securely to dry, intact skin; seals the wound margins to help prevent exudate from leaking on surrounding tissue, minimizing risk of maceration Breathable, yet moisture and bacteria resistant outer film layer Absorbent inner hydrocellular foam pad Dressing may be lifted and repositioned without losing its adherent properties Maintains a moist wound environment to help promote better healing