Learn More
Med Vet International McKesson Hydrocolloid Dressing, 4 x 4 Inch, 1887, Each
Catalog No. 503045233
Encompass_Preferred
Hydrocolloid Dressing McKesson 4 X 4 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a film backing and absorbs wound exudates The 4- by 4-inch sterile dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination The hydrocolloid film dressing has a 7-day wear time
Catalog No. 50-304-5233 Supplier Med Vet International Supplier No. 882992EA
Hydrocolloid Dressing McKesson 4 X 4 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a film backing and absorbs wound exudates The 4- by 4-inch sterile dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination The hydrocolloid film dressing has a 7-day wear time