Learn More
Med Vet International McKesson Hydrocolloid Dressing with Foam Backing, 4 x 4 Inch, 1889, Each
Catalog No. 503043095
Encompass_Preferred
Hydrocolloid Dressing McKesson 4 X 4 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The 4- x 4-inch hydrocolloid foam dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time
Catalog No. 50-304-3095 Supplier Med Vet International Supplier No. 882995EA
Hydrocolloid Dressing McKesson 4 X 4 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The 4- x 4-inch hydrocolloid foam dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time