Learn More
Med Vet International McKesson Hydrocolloid Dressing, 6 x 6 Inch, 1890, Each
Catalog No. 503045224
Encompass_Preferred
Hydrocolloid Dressing McKesson 6 X 6 Inch Square Sterile The off-white McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a foam backing and a highly absorbent inner layer that absorbs wound exudates The square sterile dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination The 6- by 6-inch foam dressing has a 7-day wear time
Catalog No. 50-304-5224 Supplier Med Vet International Supplier No. 883000EA
Hydrocolloid Dressing McKesson 6 X 6 Inch Square Sterile The off-white McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a foam backing and a highly absorbent inner layer that absorbs wound exudates The square sterile dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination The 6- by 6-inch foam dressing has a 7-day wear time