Learn More
Med Vet International McKesson Hydrocolloid Dressing, 6 x 7 Inch Sacral, 1888, 80/Case
Catalog No. 503045245
Encompass_Preferred
Hydrocolloid Dressing McKesson 6 X 7 Inch Sacral Sterile McKesson hydrocolloid dressing creates a moist environment that boosts healing Highly absorbent inner layer that absorbs wound exudates The 6- by 7-inch dressing is light beige and semi-transparent to provide visualization Outer layer is moisture resistant to prevent contamination 7-day wear time with a film backing
Catalog No. 50-304-5245 Supplier Med Vet International Supplier No. 882993CS
Hydrocolloid Dressing McKesson 6 X 7 Inch Sacral Sterile McKesson hydrocolloid dressing creates a moist environment that boosts healing Highly absorbent inner layer that absorbs wound exudates The 6- by 7-inch dressing is light beige and semi-transparent to provide visualization Outer layer is moisture resistant to prevent contamination 7-day wear time with a film backing