Learn More
Med Vet International McKesson Hydrocolloid Dressing, 2 x 2 Inch, 1882, Each
Catalog No. 503042868
Encompass_Preferred
Hydrocolloid Dressing McKesson 2 X 2 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The thin hydrocolloid dressing is semi-transparent to allow visualization of wound site The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 2 x 2 inches
Catalog No. 50-304-2868 Supplier Med Vet International Supplier No. 882980EA
Hydrocolloid Dressing McKesson 2 X 2 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The thin hydrocolloid dressing is semi-transparent to allow visualization of wound site The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 2 x 2 inches