Learn More
Med Vet International McKesson Hydrocolloid Dressing, 4 x 4 Inch, 1883, 10/Box
Catalog No. 503042869
Encompass_Preferred
Hydrocolloid Dressing McKesson 4 X 4 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudate The sterile dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination This light beige, thin hydrocolloid, 4 by 4 inch dressing has a 7-day wear time
Catalog No. 50-304-2869 Supplier Med Vet International Supplier No. 882982BX
Hydrocolloid Dressing McKesson 4 X 4 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudate The sterile dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination This light beige, thin hydrocolloid, 4 by 4 inch dressing has a 7-day wear time