Learn More
Med Vet International McKesson Hydrocolloid Dressing, 6 x 7 Inch Sacral, 1885, Each
Catalog No. 503045227
Encompass_Preferred
Hydrocolloid Dressing McKesson 6 X 7 Inch Sacral Sterile McKesson thin hydrocolloid dressing creates a moist environment that boosts healing It has a highly-absorbent inner layer that absorbs wound exudates The foam dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 6 x 7 inch sacral
Catalog No. 50-304-5227 Supplier Med Vet International Supplier No. 882986EA
Hydrocolloid Dressing McKesson 6 X 7 Inch Sacral Sterile McKesson thin hydrocolloid dressing creates a moist environment that boosts healing It has a highly-absorbent inner layer that absorbs wound exudates The foam dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 6 x 7 inch sacral