Learn More
Med Vet International McKesson Hydrocolloid Dressing, 6 x 6 Inch, 1884, Each
Catalog No. 503045200
Encompass_Preferred
Hydrocolloid Dressing McKesson 6 X 6 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The thin hydrocolloid dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 6 x 6 inches
Catalog No. 50-304-5200 Supplier Med Vet International Supplier No. 882983EA
Hydrocolloid Dressing McKesson 6 X 6 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The thin hydrocolloid dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 6 x 6 inches