Learn More
Med Vet International McKesson Hydrogel Amorphous Dressing, 1 oz., 1721, 30/Case
Catalog No. 503045346
Encompass_Preferred
Helps maintain a moist wound healing environment to assist in autolytic debridement Amorphous gel dressing comes in 1 oz. squeeze bottle
Catalog No. 50-304-5346 Supplier Med Vet International Supplier No. 949422CS
Helps maintain a moist wound healing environment to assist in autolytic debridement Amorphous gel dressing comes in 1 oz. squeeze bottle