Learn More
Med Vet International McKesson Hydrogel Gauze Dressing, 4 x 4 Inch, 61-53044, Each
Catalog No. 503044596
Encompass_Preferred
Hydrogel Gauze Dressing McKesson 4 X 4 Inch Square 1 per Pack Sterile McKesson hydrogel impregnated gauze dressing is the primary layer, protecting the wound; works well on light or no exudation areas for adequate care Sterile gauze dressing helps keep the injury clean; protect outer edge with barrier film or ointment, generating a healing environment Hydrogel maintains moisture, covering the wound, keeping the gauze soft and moist as it conforms to the area bed and helps heal the injury Impregnated wound dressing also facilitates autolytic debridement Gauze measures 4 x 4 inches, covering minor wounds
Catalog No. 50-304-4596 Supplier Med Vet International Supplier No. 488936EA
Hydrogel Gauze Dressing McKesson 4 X 4 Inch Square 1 per Pack Sterile McKesson hydrogel impregnated gauze dressing is the primary layer, protecting the wound; works well on light or no exudation areas for adequate care Sterile gauze dressing helps keep the injury clean; protect outer edge with barrier film or ointment, generating a healing environment Hydrogel maintains moisture, covering the wound, keeping the gauze soft and moist as it conforms to the area bed and helps heal the injury Impregnated wound dressing also facilitates autolytic debridement Gauze measures 4 x 4 inches, covering minor wounds