Learn More
Med Vet International McKesson Antibacterial Foam Dressing, 4 x 4 Inch, 4940, 100/Case
Catalog No. 503044893
Encompass_Preferred
Antibacterial Foam Dressing McKesson 4 X 4 Inch Square Non-Adhesive without Border Sterile Provides fast-wicking action, superior aeration and excellent absorbency Promotes healing through a moist wound environment Helps prevent recolonization Reduces the risk of maceration Conformable Waterproof and bacterial barrier PHMB (polyhexamethylene biguanide) kills and inhibits the growth of bacteria 4 x 4 inches
Catalog No. 50-304-4893 Supplier Med Vet International Supplier No. 1138632CS
Antibacterial Foam Dressing McKesson 4 X 4 Inch Square Non-Adhesive without Border Sterile Provides fast-wicking action, superior aeration and excellent absorbency Promotes healing through a moist wound environment Helps prevent recolonization Reduces the risk of maceration Conformable Waterproof and bacterial barrier PHMB (polyhexamethylene biguanide) kills and inhibits the growth of bacteria 4 x 4 inches