Learn More
Med Vet International Allevyn Life Silicone Adhesive with Border Silicone Foam Dressing, 8½ x 9 Inch, Sterile, 66801307, 10/Box
Catalog No. 503043170
Encompass_Preferred
The pink Allevyn Life foam dressings contain hydrocellular foam, a highly absorbent core and masking layers Their special shape supports use in the sacral area and fits around the cleft The masking layer keeps strike-through visibility low An absorbent core locks in fluid and reduces odor The foam layer cushions and protects with pressure redistribution A silicone contact layer minimizes wound trauma during removal Use the visual indicator to know when it's time to change an Allevyn Life sacrum dressing Sacral dressing is 21.6 x 23 centimeters
Catalog No. 50-304-3170 Supplier Med Vet International Supplier No. 839952BX
The pink Allevyn Life foam dressings contain hydrocellular foam, a highly absorbent core and masking layers Their special shape supports use in the sacral area and fits around the cleft The masking layer keeps strike-through visibility low An absorbent core locks in fluid and reduces odor The foam layer cushions and protects with pressure redistribution A silicone contact layer minimizes wound trauma during removal Use the visual indicator to know when it's time to change an Allevyn Life sacrum dressing Sacral dressing is 21.6 x 23 centimeters