Learn More
Med Vet International Allevyn Life Silicone Adhesive with Border Silicone Foam Dressing, 4 x 4 Inch, Sterile, 66801067, 100/Case
Catalog No. 503043940
Encompass_Preferred
Allevyn Life foam dressings are multi-layered with soft hydrocellular foam and a masking layer A quadrilobe shape fits curved areas of the body to keep the dressing in place when wet The masking layer reduces visibility of strike through Fluid and odor stay locked in the absorbent core The foam cushions and protects the wound by redistributing pressure A silicone contact layer minimizes wound disturbance during changes Use the visible indicator to know when it's time to switch to a fresh Allevyn Life wound dressing 10.3 x 10.3 centimeters
Catalog No. 50-304-3940 Supplier Med Vet International Supplier No. 834478CS
Allevyn Life foam dressings are multi-layered with soft hydrocellular foam and a masking layer A quadrilobe shape fits curved areas of the body to keep the dressing in place when wet The masking layer reduces visibility of strike through Fluid and odor stay locked in the absorbent core The foam cushions and protects the wound by redistributing pressure A silicone contact layer minimizes wound disturbance during changes Use the visible indicator to know when it's time to switch to a fresh Allevyn Life wound dressing 10.3 x 10.3 centimeters