Learn More
Med Vet International Thin Foam Dressing Mepilex Lite 4 X 4 Inch Without Border Film Backing Silicone Adhesive Square Sterile, 284190, 5/Box
Catalog No. 503043953
Encompass_Preferred
Mepilex Lite is a thin and highly conformable foam dressing that absorbs exudate and maintains a moist wound environment The Safetac layer seals around the wound edges which prevents exudate from leaking onto the surrounding skin, thus minimizing the risk for maceration Promotes patient comfort; thin and very comfortable to wear May be left in place during radiation therapy using commonly applied energy ranges and absorbed doses Well suited to be used under compression bandages May be cut to suit various wound shapes and difficult-to-dress locations
Catalog No. 50-304-3953 Supplier Med Vet International Supplier No. 563657BX
Mepilex Lite is a thin and highly conformable foam dressing that absorbs exudate and maintains a moist wound environment The Safetac layer seals around the wound edges which prevents exudate from leaking onto the surrounding skin, thus minimizing the risk for maceration Promotes patient comfort; thin and very comfortable to wear May be left in place during radiation therapy using commonly applied energy ranges and absorbed doses Well suited to be used under compression bandages May be cut to suit various wound shapes and difficult-to-dress locations