Learn More
Med Vet International McKesson Wound Protector, Small, 7 X 7 Inch, 11077, 5/Pack
Catalog No. 503044146
Encompass_Preferred
Wound Protector McKesson Small Adhesive Helps keep sites, wounds and dressings dry while the patient showers Protects surgical sites, intravenous (IV) sites, catheters, PICC (peripherally inserted central catheter) lines, stomas, ostomies, and primary dressings Not intended for use in the bath or for swimming; should not be submerged Water resistant 7 x 7 inches
Catalog No. 50-304-4146 Supplier Med Vet International Supplier No. 1162466PK
Wound Protector McKesson Small Adhesive Helps keep sites, wounds and dressings dry while the patient showers Protects surgical sites, intravenous (IV) sites, catheters, PICC (peripherally inserted central catheter) lines, stomas, ostomies, and primary dressings Not intended for use in the bath or for swimming; should not be submerged Water resistant 7 x 7 inches