Learn More
Med Vet International Smith & Nephew Exu-Dry Anti-Shear Super Absorbent Dressing, 4 x 6 inch, Sterile, 5999004120, 10/Box
Catalog No. 503043116
Encompass_Preferred
Exu-Dry is a one-piece wound dressing made up of multiple layers Exu-Dry's anti-shear layer helps reduce shearing thus may help protect fragile granulation tissue Highly absorbent, Exu-Dry helps reduce the risk of maceration and irritation to the wound and surrounding tissue Exu-Dry wicks away drainage from the wound into the entire dressing, reducing the frequency of dressing changes Because Exu-Dry is permeable and non-occlusive, it can be used as a wet or dry dressing on clean or contaminated wounds Compatible with most topical agents, liquids and creams Full absorbency Nonadherent to the wound, Exu-Dry is more comfortable for patients during and between dressing changes 10 x 15 centimeters
Catalog No. 50-304-3116 Supplier Med Vet International Supplier No. 388241BX
Exu-Dry is a one-piece wound dressing made up of multiple layers Exu-Dry's anti-shear layer helps reduce shearing thus may help protect fragile granulation tissue Highly absorbent, Exu-Dry helps reduce the risk of maceration and irritation to the wound and surrounding tissue Exu-Dry wicks away drainage from the wound into the entire dressing, reducing the frequency of dressing changes Because Exu-Dry is permeable and non-occlusive, it can be used as a wet or dry dressing on clean or contaminated wounds Compatible with most topical agents, liquids and creams Full absorbency Nonadherent to the wound, Exu-Dry is more comfortable for patients during and between dressing changes 10 x 15 centimeters