Learn More
Med Vet International DermaRite Oil Emulsion Impregnated Dressing, 3 x 8 Inch, Sterile, 22380, Each
Catalog No. 503044916
Encompass_Preferred
Oil emulsion wound dressing consists of a nonadherent gauze mesh impregnated with white petrolatum in an oil emulsion blend This primary dressing permits the flow of exudate without sticking to the wound bed Conforms to all body contours Nonadherent, use beneath a secondary dressing to manage exudate and securement
Catalog No. 50-304-4916 Supplier Med Vet International Supplier No. 1123829EA
Oil emulsion wound dressing consists of a nonadherent gauze mesh impregnated with white petrolatum in an oil emulsion blend This primary dressing permits the flow of exudate without sticking to the wound bed Conforms to all body contours Nonadherent, use beneath a secondary dressing to manage exudate and securement