Learn More
Med Vet International Xeroform Petrolatum Impregnated Dressing, Rectangle, 5 X 9 Inch, Sterile, 24590, 50/Box
Catalog No. 503045117
Encompass_Preferred
The sterile Xeroform gauze dressing helps maintain a conducive wound healing environment for faster recovery It combines petrolatum and bismuth tribromophenate to manage wounds with zero to minimal fluid discharge This nonadherent gauze dressing won't stick to the skin, minimizing pain during removal Dressing adapts to body contours to ensure proper wound sealing and prevent contamination 5 x 9 inches
Catalog No. 50-304-5117 Supplier Med Vet International Supplier No. 946777BX
 Restricted Item
Add to Cart
Add to Cart
The sterile Xeroform gauze dressing helps maintain a conducive wound healing environment for faster recovery It combines petrolatum and bismuth tribromophenate to manage wounds with zero to minimal fluid discharge This nonadherent gauze dressing won't stick to the skin, minimizing pain during removal Dressing adapts to body contours to ensure proper wound sealing and prevent contamination 5 x 9 inches