Learn More
Med Vet International Xeroform Petrolatum Impregnated Dressing, Rectangle, 5 X 9 Inch, Sterile, 24590, Each
Catalog No. 503043076
Encompass_Preferred
The sterile Xeroform gauze dressing helps maintain a conducive wound healing environment for faster recovery It combines petrolatum and bismuth tribromophenate to manage wounds with zero to minimal fluid discharge This nonadherent gauze dressing won't stick to the skin, minimizing pain during removal Dressing adapts to body contours to ensure proper wound sealing and prevent contamination 5 x 9 inches
Catalog No. 50-304-3076 Supplier Med Vet International Supplier No. 946777EA
The sterile Xeroform gauze dressing helps maintain a conducive wound healing environment for faster recovery It combines petrolatum and bismuth tribromophenate to manage wounds with zero to minimal fluid discharge This nonadherent gauze dressing won't stick to the skin, minimizing pain during removal Dressing adapts to body contours to ensure proper wound sealing and prevent contamination 5 x 9 inches