Learn More
Med Vet International Foam Dressing Proximel 6 X 6 Inch With Border Waterproof Film Backing Silicone Adhesive Square Sterile, 14400000, Each
Catalog No. 503044394
Encompass_Preferred
Silicone adhesive is gentle on patients' skin to reduce irritation and pain, enabling the dressing to be changed without stripping epidermal cells Can be lifted or repositioned without adherence failure Dressing core supports fast absorption and helps minimize maceration risk High breathability (MVTR) supports good moisture balance Waterproof, bacteria proof and air-permeable
Catalog No. 50-304-4394 Supplier Med Vet International Supplier No. 1242034EA
Silicone adhesive is gentle on patients' skin to reduce irritation and pain, enabling the dressing to be changed without stripping epidermal cells Can be lifted or repositioned without adherence failure Dressing core supports fast absorption and helps minimize maceration risk High breathability (MVTR) supports good moisture balance Waterproof, bacteria proof and air-permeable