Learn More
Med Vet International Proximel Silicone Adhesive Foam Dressing, 9.2 x 9.2 Inch, Sterile, 14700000, 50/Case
Catalog No. 503043342
Encompass_Preferred
Proximel silicone foam dressing stays in place during routine skin or wound checks while promoting healing and supporting prevention protocols Reduces and redistributes pressure across the surface to reduce both peak and overall level of force Proximel's distinctive properties allow you to confidently lift and reposition the dressing while reducing the risk of adherence failure High MVTR rate supports good moisture balance Superabsorbent core supports fast absorption and helps minimize maceration risk 9.2 x 9.2 inches
Catalog No. 50-304-3342 Supplier Med Vet International Supplier No. 1242734CS
Proximel silicone foam dressing stays in place during routine skin or wound checks while promoting healing and supporting prevention protocols Reduces and redistributes pressure across the surface to reduce both peak and overall level of force Proximel's distinctive properties allow you to confidently lift and reposition the dressing while reducing the risk of adherence failure High MVTR rate supports good moisture balance Superabsorbent core supports fast absorption and helps minimize maceration risk 9.2 x 9.2 inches