Learn More
Med Vet International Silver Tracheostomy Dressing Algidex Ag 4 X 4 Inch, 46-TD44, 10/Case
Catalog No. 503044307
Encompass_Preferred
Pre-cut trach dressing Unique combination of silver, alginate, and maltodextrin designed to create the optimal moist wound environment important to healing Excellent exudate management, helping reduce the risk of peri-wound maceration Will not adhere to wound upon removal
Catalog No. 50-304-4307 Supplier Med Vet International Supplier No. 719734CS
Pre-cut trach dressing Unique combination of silver, alginate, and maltodextrin designed to create the optimal moist wound environment important to healing Excellent exudate management, helping reduce the risk of peri-wound maceration Will not adhere to wound upon removal