Learn More
Med Vet International McKesson Octagonal Sterile Dressing with Frame-Style Delivery, 2-3/8 x 2-3/4 Inch, Transparent, 4985, 400/Case
Catalog No. 503043702
Encompass_Preferred
Transparent Film Dressing McKesson Octagon 2-3/8 X 2-3/4 Inch Frame Style Delivery Without Label Sterile Sterile dressing is semi-permeable and water resistant Intended to provide a moist environment for granulating or superficial wounds Transparent film allows the site to be visually monitored Intended for single-use applications
Catalog No. 50-304-3702 Supplier Med Vet International Supplier No. 886408CS
Transparent Film Dressing McKesson Octagon 2-3/8 X 2-3/4 Inch Frame Style Delivery Without Label Sterile Sterile dressing is semi-permeable and water resistant Intended to provide a moist environment for granulating or superficial wounds Transparent film allows the site to be visually monitored Intended for single-use applications