Learn More
Med Vet International Super Absorbent Dressing Zetuvit Plus 4 X 4 Inch Square, 413110, 10/Box
Catalog No. 503044491
Encompass_Preferred
Soft wound contact layer ensures fast absorption of wound exudate into the dressing and prevents adherence to the wound Quick and uniform distribution of exudate into the superabsorbent core Very high absorption capacity due to highly absorbent core High wearing comfort due to excellent padding properties Water repellent outer later protects clothing and bedding
Catalog No. 50-304-4491 Supplier Med Vet International Supplier No. 1161228BX
Soft wound contact layer ensures fast absorption of wound exudate into the dressing and prevents adherence to the wound Quick and uniform distribution of exudate into the superabsorbent core Very high absorption capacity due to highly absorbent core High wearing comfort due to excellent padding properties Water repellent outer later protects clothing and bedding