Learn More
Med Vet International DermaGauze Impregnated Dressing, 4 x 4 Inch, Sterile, 00250E, Each
Catalog No. 503044276
Encompass_Preferred
Has the ability to cool the wound surface and due to its high moisture content, protect exposed nerve surfaces Nonadherent to tissue Provides an optimal moist wound environment for healing Prevents damage to healthy tissue
Catalog No. 50-304-4276 Supplier Med Vet International Supplier No. 727081EA
Has the ability to cool the wound surface and due to its high moisture content, protect exposed nerve surfaces Nonadherent to tissue Provides an optimal moist wound environment for healing Prevents damage to healthy tissue