Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Drug and Poison Analysis: Chemistry and Toxicology Educational Kit
GREENER_CHOICESCI_ED

Catalog No. s28501 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S28501 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S28501 Supplier Innovating Science™ Supplier No. IS9016
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Phenolphthalein/ Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer and reproductive harm.

Includes

10g Aspirin; 10g Acetaminophen; 10g Sodium Bicarbonate Antacid; 10g Effervescent Antacid; 10g Unknown #1; 10g Unknown #2; 30mL Universal Indicator Solution; 30mL 0.5M Hydrochloric Acid; 30mL Ferric Nitrate; 1g Simulated Marijuana; 10g Simulated LSD; 10g Simulated Cocaine; 30mL Simulated Scott Reagent; 30mL Simulated Van Urk Reagent; 30mL Simulated Lead Solution; 30mL Simulated Mercury Solution; 30mL Unknown Poison; 30mL Simulated Potassium Chromate; 30mL Simulated Urine Sample; 30mL Alanine Control; 30mL Glycine Control Spray Bottle w/0.5g Ninhydrin Powder; 4 x 25mL 95% Ethanol; 15 x 25ml Chromatography Solvent; 15 Universal Indicator Color Charts; 6 Capillary Tubes; 15pc Chromatography Sheets; 30 Spot Plates; 8 Scoops; 1pkg Toothpicks

To learn the difference between forensic chemistry and forensic toxicology. Innovating Science Forensic Drug and Poison Analysis: Chemistry and Toxicology Educational Kit, helps students to perform presumptive chemical tests on drugs and poisons, identify unknown samples based on chemical test results and examine urine for evidence of heavy metal poisoning using chromatography.

  • Experiments to understand how over-the-counter drugs, controlled drugs, and illegal drugs may act as toxins
  • Contains enough material for 15 groups
  • Teacher’s Manual and Student Study Guide copymasters are included

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational use
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 15 Lab Groups
Product Type Forensic Drug and Poison Analysis: Chemistry and Toxicology Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.