Description
- Features a pupil gauge 1.0 to 9.0 mm visible from both sides and provides a quick reference for physical reactions to drugs.
- Features a sturdy, black leatherette case.
- Made from translucent plastic and leatherette.
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Not Applicable |
| Detectable Analytes | CNS Depressants, CNS Stimulants, Hallucinogens, Phencyclidine (PCP), Narcotics, Inhalants |
| DoA Calibrators | Not Applicable |
| Form | Recognition Card |