Learn More
Selleck Chemical LLC FDA-approved & Passed Phase I Drug Library 100uL/well
SDP
Catalog No. 501948382
Encompass_Preferred
A unique collection of 3176 drugs that are marketed around the world or have passed clinical phase 1 and can be used for high throughput screening (HTS) and high content screening (HCS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8382 Supplier Selleck Chemical LLC Supplier No. L3800100UL/WELL
Add to Cart
Add to Cart
Request a Quote
A unique collection of 3176 drugs that are marketed around the world or have passed clinical phase 1 and can be used for high throughput screening (HTS) and high content screening (HCS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.