Learn More
Alkali Scientific Drug Testing Container with Graduations, Polypropylene, Attached White Screw Cap, Tamper Seal, Patient ID Label & Temperature Strip, 95 kPa rated, Sterile, 60mL, 500/Case
SDP
Catalog No. 5000662819
Encompass_Preferred
Urine Drug Testing Containers from Alkali Scientific are designed to support secure, compliant specimen collection and transport in clinical and occupational health environments. Manufactured from durable polypropylene, each container features an attached white screw cap with an integrated tamper evident seal to help maintain specimen integrity and support chain of custody requirements. A built in temperature strip allows immediate specimen validation at the time of collection, while the included patient ID label supports accurate identification and documentation. Each container is 95 kPa rated to provide reliable, leak resistant performance during handling and transport, including air shipment. Suitable for use in DOT and non DOT testing programs, these containers support consistent specimen collection workflows.
Catalog No. 50-006-62819 Supplier Alkali Scientific Supplier No. DTC060
Urine Drug Testing Containers from Alkali Scientific are designed to support secure, compliant specimen collection and transport in clinical and occupational health environments. Manufactured from durable polypropylene, each container features an attached white screw cap with an integrated tamper evident seal to help maintain specimen integrity and support chain of custody requirements. A built in temperature strip allows immediate specimen validation at the time of collection, while the included patient ID label supports accurate identification and documentation. Each container is 95 kPa rated to provide reliable, leak resistant performance during handling and transport, including air shipment. Suitable for use in DOT and non DOT testing programs, these containers support consistent specimen collection workflows.