Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Drum Cover

Catalog No. 19046158 Shop All Justrite Safety Group Products
Change view
Click to view available options
Description:
Snap-on
Diameter (English) Inner:
18.37 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Diameter (English) Inner
19-046-158 Snap-on 18.37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-046-158 Supplier Justrite™ Supplier No. 26730
  • $404.00
    $240.65 / Each of 1

    Save $163.35

    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convert open-top 30 and 55 gal. (114 and 208L) drums into fire-safe receptacles

Self-closing cover inexpensively converts steel drums into fire-safe receptacles for combustible trash.

  • Requires screwdriver and wrench for easy attachment and removal to drum
  • Internal replaceable fusible link melts at 74°C (165°F), automatically closing cover in the event of a fire
  • Highly chemical-resistant powder-coat paint finish

Certifications

FM approved.

Specifications

Certifications/Compliance FM, TUV
Product Type Drum Cover
Description Snap-on
Color Red
For Use With (Application) Convert open-top 30 gal. drums into fire-safe receptacles
For Use With (Equipment) 30 gal. Drums
Material Steel
Includes Internal Replaceable Fusible Link, Hardware
Diameter (English) Inner 18.37 in.
Diameter (Metric) Inner 46.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.