Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Drum Grate Replacement for EcoPolyBlend™ Spill Pallets and Accumulation Center

Catalog No. 19822460 Shop All Justrite Safety Group Products
Change view
Click to view available options
Dimensions (L x W x H):
24 x 24 x 2.5 in.
48 x 24 x 2.5 in.
For Use With (Equipment):
2-Drum Centers
3 In-Line or 1-Drum Centers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) For Use With (Equipment)
19-822-460 48 x 24 x 2.5 in. 2-Drum Centers
19-822-459 24 x 24 x 2.5 in. 3 In-Line or 1-Drum Centers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-460 Supplier Justrite™ Supplier No. 28259
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bring old equipment back into service with replacement parts. The Justrite™ Drum Grate Replacement for EcoPolyBlend™ Spill Pallets and Accumulation Center is a replacement replacement polyethylene grating.

  • Justrite’s steel laboratory safety cans have all the benefits of the Type I cans
  • Added features make these the choice of lab professionals for safely dispensing hazardous liquids into test tubes, small flasks and beakers
  • Fill spouts have self-close, pressure-relief caps and stainless steel flame arresters
  • These cans also feature a vacuum-relief valve within the lid for smooth liquid flow, and self-closing faucets for trouble-free dispensing

Specifications

Color Black
Description Recycled Polyethylene
Material Polyethylene
For Use With (Equipment) 2-Drum Centers
Dimensions (L x W x H) 48 x 24 x 2.5 in.
Width (English) 24 in.
Height (English) 2.5 in.
Depth (English) 48 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.