Learn More
BURKLE INC Drum holder for placing filling containers, V2A
Supplier:  BURKLE INC 56180200
Encompass_Preferred
The height-adjustable support surface of the barrel holder allows the filling container to be placed directly under the outlet of the barrel pump. This eliminates the risk of spillage when filling from a great height, such as when the container is on the floor. It also allows the operator to have both hands free while filling and to easily remove larger quantities. A scale with container can also be placed on the support surface to accurately measure the liquid to be filled.
The holder can be used for Bürkle pumps with discharge elbow or discharge hose. Discharge hoses can be easily inserted into the hinged holder. Suitable for all plastic and stainless steel drums from 60 liters to 220 liters with a minimum rim height of 10 mm.
Catalog No. NC2829126