Learn More
Ampco Drum Opener
SDP
Catalog No. 19173597 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
11 -7/8 in.
Catalog No. Length (English)
19-173-597 11 -7/8 in.
1 options
Catalog No. 19-173-597 Supplier Ampco Supplier No. DO100
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant drum opener.
  • Please see image for additional information regarding measurements.
  • Tip width (a): 2 and 3/8 in.
  • Tip length (b): 3 in.
  • Overall length: 11 and 7/8 in.
Specifications
Product Type Drum Opener
Length (English) 11 -7/8 in.
Length (English) Blade 3 in.
Quantity 1 Ea.