Learn More
Grainger DAYTON Battery Operated Pail Pump
Catalog No. NC1274345
Encompass_Preferred
The Dayton Battery Operated Pail Pump is a portable, battery-powered liquid transfer pump designed for light-duty fluid transfer applications. It features a polypropylene housing, thermoplastic elastomer pump, and stainless steel fasteners for corrosion resistance. This pump has a 1 GPM flow rate and will pump up to 5 feet vertically. It runs on 4 C alkaline batteries (not included) and has an on/off switch located on the handle for convenient operation. The pump's design allows it to be fully submerged during operation. Overall, this is a lightweight, portable pump well-suited for transferring water and light fluids in applications with moderate flow rate requirements.
Catalog No. NC1274345 Supplier Grainger Supplier No. 38Y794
The Dayton Battery Operated Pail Pump is a portable, battery-powered liquid transfer pump designed for light-duty fluid transfer applications. It features a polypropylene housing, thermoplastic elastomer pump, and stainless steel fasteners for corrosion resistance. This pump has a 1 GPM flow rate and will pump up to 5 feet vertically. It runs on 4 C alkaline batteries (not included) and has an on/off switch located on the handle for convenient operation. The pump's design allows it to be fully submerged during operation. Overall, this is a lightweight, portable pump well-suited for transferring water and light fluids in applications with moderate flow rate requirements.