Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Drum Pump Tube 16 L 316 SS 1-1/4 I.D.

Catalog No. 502756070
Encompass_Preferred

Drum Pump Tube 16 L 316 SS 1-1/4 I.D.

Catalog No. 50-275-6070 Supplier Grainger Supplier No. 15R523
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Drum Pump Tube 16 L 316 SS 1-1/4 I.D.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.