Learn More
MSC Milwaukee Tool Replacement Drum and Push Cable, 85 ft, 1.5 in. to 4 in. Drain Lines, 1 ea
Catalog No. 5000510620
Encompass
This replacement drum and cable is designed for use with the M12™ 85 ft. High Flex Compact Inspection Camera. It allows for easy attachment to the powered base via a quick-release knob and enables navigation through tight bends and traps in 1.5 in. to 4 in. drain lines up to 85 ft. long.
  • Provides a balance of push and flexibility for fixture-to-branch line inspections
  • Supports inspection of 1.5 in. to 4 in. diameter drain lines
  • Includes quick-release knob for simple drum connection
  • Compatible with 85 ft. length requirements
Catalog No. 50-005-10620 Supplier MSC Supplier No. 72156359
This replacement drum and cable is designed for use with the M12™ 85 ft. High Flex Compact Inspection Camera. It allows for easy attachment to the powered base via a quick-release knob and enables navigation through tight bends and traps in 1.5 in. to 4 in. drain lines up to 85 ft. long.
  • Provides a balance of push and flexibility for fixture-to-branch line inspections
  • Supports inspection of 1.5 in. to 4 in. diameter drain lines
  • Includes quick-release knob for simple drum connection
  • Compatible with 85 ft. length requirements