Learn More
CONTROLS FOR AUTOMATION DrumQuik PRO, Blue, Glass-filled Nylon
Catalog No. NC1479720
Encompass
DrumQuik PRO systems create a closed transfer system for general chemicals. Rugged and cost-effective DrumQuik dispensing systems minimize chemical exposure and facilitate the delivery of chemicals from bulk transfer containers to end use applications.
Catalog No. NC1479720 Supplier CONTROLS FOR AUTOMATION Supplier No. 2408900
DrumQuik PRO systems create a closed transfer system for general chemicals. Rugged and cost-effective DrumQuik dispensing systems minimize chemical exposure and facilitate the delivery of chemicals from bulk transfer containers to end use applications.