Learn More
Uline DRUM SEALS - 12"""", BLUE, 100 pack
Catalog No. NC1921900
Encompass
Tug-Tight Drum Seals - 12", BlueBreaking strength is 50 lbs.Pulls tight - Lids stay locked on.Consecutively numbered for easy tracking.
Catalog No. NC1921900 Supplier Uline Supplier No. H610BLU
Tug-Tight Drum Seals - 12", BlueBreaking strength is 50 lbs.Pulls tight - Lids stay locked on.Consecutively numbered for easy tracking.