Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JUSTRITE Drum Storage Cabinets, 55 gal., Self-Close, with Drum Support

Catalog No. 18666739 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Blue
Gray
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-666-739 Yellow
18-666-740 Red
18-666-741 Blue
18-666-742 Gray
18-666-743 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-666-739 Supplier JUSTRITE Supplier No. 896220
Add to Cart
Add to Cart

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

Width (Metric) Interior 864 mm
Width (Metric) Exterior 781 mm
Weight (English) 347 lb.
Weight (Metric) 157.4 kg
Width (English) Interior 30.8 in.
Load Bearing Capacity 350 lb./shelf
No. of Shelves 1
No. of Doors 2
Surface Coating Powder-coat Paint
Height (Metric) Exterior 1651 mm
Height (Metric) Interior 1537 mm
Explosion Relief Panel No
Height (English) Interior 60.5 in.
Door Style Self-close, Locking, Three-point lock
Width (English) Exterior 34 in.
Product Type Vertical Drum Storage Cabinet
Material Steel (18 ga), Galvanized Steel
Dimensions (D x W x H) 34 x 34 x 65 in.
Color Yellow
Depth (English) Exterior 34 in.
Height (English) Exterior 65 in.
Dimensions (D x W x H) Interior 30.3 x 30.8 x 60.5 in.
Depth (Metric) Interior 771 mm
Depth (Metric) Exterior 864 mm
Depth (English) Interior 30.3 in.
Certifications/Compliance FM Approved
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.