Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Drum Storage Cabinets
GREENER_CHOICE

Catalog No. 17987415 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Dimensions (D x W x H) Interior:
28.25 x 28.25 x 62 in.
28.5 x 55 x 62 in.
Width (English) Exterior:
31.25 in.
58 in.
Weight (English):
334 lb.
344 lb.
575 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior Width (English) Exterior Weight (English)
17-987-415 28.25 x 28.25 x 62 in. 31.25 in. 344 lb.
18-887C 28.5 x 55 x 62 in. 58 in. 575 lb.
17-156-1A 28.25 x 28.25 x 62 in. 31.25 in. 334 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-987-415 Supplier Eagle™ Supplier No. 2610X
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Drum supports and built-in grounding connectors. Cabinets for hazardous waste drums include drum rollers for easy drum handling.

The Eagle Drum Storage Cabinet is designed to let you store 55-gallon drums containing hazardous waste or flammables.

  • Safely store hazardous waste or flammables in the workplace
  • One- and two-drum capacity with choice of manual or self-closing doors
  • Store drums vertically or horizontally
  • Vertical cabinets include half shelf for additional storage of accessories or containers
  • Cabinets feature large red FLAMMABLE - KEEP FIRE AWAY and HAZARDOUS WASTE labels on yellow enamel finish; flammables cabinets have FLAMMABLE - KEEP FIRE AWAY label

Certifications

FM approved (except 17-516-1B).

Compliance

Comply with NFPA Code 30, and meet OSHA 29CFR1910.106(d)3 standard.

‡Accommodates no more than 60 gal. (227L) may be Class I and Class II liquids.

†Not FM approved.

Specifications

Dimensions (D x W x H) Interior 28.25 x 28.25 x 62 in.
Load Bearing Capacity 55 gal.
Product Type Vertical drum storage cabinet
Depth (Metric) Interior 71.75 cm
Depth (Metric) Exterior 79.37 cm
Weight (Metric) 156.03 kg
Width (Metric) Exterior 79.3 cm
Height (Metric) Exterior 165.1 cm
Width (Metric) Interior 71.75 cm
Height (Metric) Interior 157.4 cm
Description One drum, vertical, two door, self-close doors
No. of Shelves 1
No. of Doors 2
Weight (English) 344 lb.
Color Yellow
For Use With (Application) Flammables
Depth (English) Exterior 31.25 in.
Height (English) Exterior 65 in.
Height (English) Interior 62 in.
Material Steel
Door Style Manual-close
Certifications/Compliance FM, NFPA, OSHA
Depth (English) Interior 28.25 in.
Width (English) Exterior 31.25 in.
Includes Drum supports and built-in grounding connectors and one shelf, large flammable keep fire away label
Width (English) Interior 28.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.