Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drummond™ Aqua-Cap Special Sample Collection Tubes

Catalog No. 13681373
Change view
Click to view available options
Volume (Metric):
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-681-373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-681-373 Supplier Drummond™ Supplier No. 70001000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Material Glass
For Use With (Application) For capillary pipets with hydrophobic plug preset to precise volume delivery
Color Blue
Volume (Metric) 100 μL
Type Sample Collection Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.