Learn More
Drummond™ Hematocrit Tubes
GSA_VA
Catalog No. 2117123
Change view
Click to view available options
Product Type:
Hematocrit Tube
Catalog No. Product Type
21-171-23 Hematocrit Tube
1 options
This item is not returnable. View return policy
Catalog No. 21-171-23 Supplier Drummond™ Supplier No. 80007520/5
Add to Cart
Add to Cart
This item is not returnable. View return policy
Drummond Polycarbonate Hematocrit Tubes provide the identical capillary action as Glass hematocrit tubes. Tube Size: 75 mm.
Drummond Hematocrit Tubes, Polycarbonate, Plain Hematocrit Tube, Certifications/Compliance: FDA, NIOSH, CDC, OSHA, For Use With: Provide the identical capillary action as Glass hematocrit tubes, Tube Size: 75mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Hematocrit Tube
Material Polycarbonate
For Use With (Application) Provide the identical capillary action as Glass hematocrit tubes
Length (Metric) 75 mm
Certifications/Compliance FDA, NIOSH, CDC, OSHA