Learn More
Drummond™ Original Portable Pipet-Aid™ Pipet Controller
GSA_VA
Catalog No. 1368108
Change view
Click to view available options
Voltage:
110 V
220 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
13-681-08 220 V
13-681-19 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 13-681-08 Supplier Drummond™ Supplier No. 4000200
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Convenient alternative to mouth pipetting in the laboratory.
  • Easy-to-use volume control mechanism permits precise fluid intake and delivery
  • Check valve protects against accidental fluid intake into the handle
  • Can be used with glass or plastic pipets
  • Autoclavable components
Order Info
Contact your Fisher Scientific Customer Service Representative for available replacement parts.
Certifications
CE, Euro version
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Autoclavable Autoclavable
Type Portable Filler/Dispenser
For Use With (Application) Laboratory pipetting
For Use With (Equipment) Glass or plastic pipets
Includes Pipet Controller, TC Nosepiece
Voltage 220 V