Learn More
Drummond™ PCR Micropipets
GSA_VA
Catalog No. 21179A
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
5 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-179A 5 μL
21-179B 1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-179A Supplier Drummond™ Supplier No. 50001001X
Add to Cart
Add to Cart
Disposable, positive displacement micropipets with stainless steel plungers
  • Measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
  • Decontaminate at temperature of 400°F or higher
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Application) To measure true sample volumes regardless of viscosity, eliminates contamination from residual aerosols
Includes Stainless Steel plungers
Volume (Metric) 5 μL
Type PCR Micropipet
Disposable Single-use