Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drummond™ Pipet-Aid Hood Mate Pipet Controller Accessories

Catalog No. p-48350
Change view
Click to view available options
Type:
Backing Plate with Tape
Coiled Hose
Tape
Valve Body Assembly
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-681-65 Backing Plate with Tape
13-681-66 Tape
13-681-67 Valve Body Assembly
13-681-68 Coiled Hose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-681-65 Supplier Drummond™ Supplier No. 4000082
Only null left
Add to Cart
Add to Cart

Specifications

Autoclavable Not Autoclavable
Color Gray
Type Backing Plate with Tape
For Use With (Equipment) Pipet-Aid™ hood mate 300 filler/dispensers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.