Learn More
Drummond™ Portable Pipet-Aid™ Pipette Controller Accessory, TC Rubber Gasket
Catalog No. 1368130
Change view
Click to view available options
TC gasket
Catalog No.
13-681-30 TC gasket
1 options
Catalog No. 13-681-30 Supplier Drummond™ Supplier No. 4000057
Add to Cart
Edge
Add to Cart
To ensure quality, replace your product's components with manufacturer's parts. Drummond TC rubber gaskets come in pairs and can be used with any of the Portable Pipet-Aid Pipette Controllers.
  • Two gaskets per pack
  • Rubber
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Original Portable Pipet-Aid, Portable Pipet-Aid XL, Portable Pipet-Aid XP