Learn More
Drummond™ Pipet-Aid™ Series Pipet Controller Accessory, EZ Grip TC Nosepiece Kit
GSA_VA
Catalog No. 1368118A
Change view
Click to view available options
For AC-powered models 13-681-15, -15C, -15D, -16; and portable model 13-681-19
Catalog No.
13-681-18A For AC-powered models 13-681-15, -15C, -15D, -16; and portable model 13-681-19
1 options
Catalog No. 13-681-18A Supplier Drummond™ Supplier No. 4000050
Add to Cart
Add to Cart
For use with Pipet-Aid™ Series Pipet Controller. Drummond™ Pipet-Aid™ Series Pipet Controller Accessory, EZ Grip TC Nosepiece Kit with four self-locking filters eliminates possibility of cross-contamination due to overpipetting.
  • Eliminates possibility of cross-contamination due to over-pipetting
  • Incorporates dual hydrophilic 0.8μm, hydrophobic 0.8μm filter which prevents further use once contaminated by fluid
  • Double membrane filter provides barrier to particles as small as 0.1μm
  • Each kit contains all necessary parts including four filters
  • Extra filters may be ordered separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Original Pipet-Aid, Original Portable Pipet-Aid
Includes Four 1 in. dia. filters, as supplied with 13-681-15E, -19, 13-681-15C and -15D, adapts any Pipet-Aid™ filler/dispenser to eliminate cross-contamination due to overpipetting